Merck
  • Stereoselective ring opening of meso bicyclic hydrazines: a straightforward approach to hydrazino cyclopentenic cores.
Stereoselective ring opening of meso bicyclic hydrazines: a straightforward approach to hydrazino cyclopentenic cores.
Organic letters (2003-12-05)
Alejandro Pérez Luna, Michèle Cesario, Martine Bonin, Laurent Micouin
ABSTRACT
The diastereoselective synthesis of hydrazinocyclopentenes 6 or 7 can be achieved in a straightforward manner from Diels-Alder adduct 4 using an acid-catalyzed rearrangement or a palladium-catalyzed allylic substitution reaction. In the latter case, enantioenriched compounds with ee values up to 58% can be obtained when an appropriate chiral ligand is used. [reaction: see text]
Sigma-Aldrich
DPPF, 97%