Merck
Pharmacokinetic/Pharmacodynamic Modeling of Tulathromycin against
Frontiers in pharmacology (2017-07-14)
Qiaoyi Zhou, Guijun Zhang, Qin Wang, Wenguang Liu, Yan Huang, Pengling Yu, Yanqin Li, Huanzhong Ding, Binghu Fang
ABSTRACT
Tulathromycin, a macrolide antibiotic, is used for the treatment of respiratory disease in cattle and swine. The aim of our study was to investigate the
Sigma-Aldrich
Tulathromycin A, ≥95% (HPLC)