Merck
  • Anti-inflammatory activity of immunoglobulin G resulting from Fc sialylation.
Anti-inflammatory activity of immunoglobulin G resulting from Fc sialylation.
Science (New York, N.Y.) (2006-08-05)
Yoshikatsu Kaneko, Falk Nimmerjahn, Jeffrey V Ravetch
ABSTRACT
Immunoglobulin G (IgG) mediates pro- and anti-inflammatory activities through the engagement of its Fc fragment (Fc) with distinct Fcg receptors (FcgRs). One class of Fc-FcgR interactions generates pro-inflammatory effects of immune complexes and cytotoxic antibodies. In contrast, therapeutic intravenous gamma globulin and its Fc fragments are anti-inflammatory. We show here that these distinct properties of the IgG Fc result from differential sialylation of the Fc core polysaccharide. IgG acquires anti-inflammatory properties upon Fc sialylation, which is reduced upon the induction of an antigen-specific immune response. This differential sialylation may provide a switch from innate anti-inflammatory activity in the steady state to generating adaptive pro-inflammatory effects upon antigenic challenge.
Sigma-Aldrich
Beta-galactoside alpha-2,6-sialyltransferase 1, ≥300 units/mg protein, ST6GAL1 human recombinant, expressed in HEK 293 cells