Merck
  • Acquired Resistance to FGFR Inhibitor in Diffuse-Type Gastric Cancer through an AKT-Independent PKC-Mediated Phosphorylation of GSK3β.
Acquired Resistance to FGFR Inhibitor in Diffuse-Type Gastric Cancer through an AKT-Independent PKC-Mediated Phosphorylation of GSK3β.
Molecular cancer therapeutics (2017-10-06)
Wen Min Lau, Eileen Teng, Kie Kyon Huang, Jin Wei Tan, Kakoli Das, Zhijiang Zang, Tania Chia, Ming Teh, Koji Kono, Wei Peng Yong, Asim Shabbir, Amy Tay, Niam Sin Phua, Patrick Tan, Shing Leng Chan, Jimmy Bok Yan So
ABSTRACT
Preclinical models of diffuse-type gastric cancer (DGC) that reliably predict clinical activity of novel compounds are lacking. To overcome the problem of poor tumor cellularity in DGC, we used cells from malignant ascites to establish DGC patient-derived xenograft (PDX) models that recapitulate the primary cancer. Cells in PDX model GAGA6 with
Sigma-Aldrich
MISSION® esiRNA, targeting human FGFR2
Sigma-Aldrich
1-Azakenpaullone, ≥97% (HPLC)
Sigma-Aldrich
Anti-β-Actin antibody, Mouse monoclonal, clone AC-15, purified from hybridoma cell culture