Merck
  • PTFE-based microreactor system for the continuous synthesis of full-visible-spectrum emitting cesium lead halide perovskite nanocrystals.
PTFE-based microreactor system for the continuous synthesis of full-visible-spectrum emitting cesium lead halide perovskite nanocrystals.
Beilstein journal of nanotechnology (2017-12-21)
Chengxi Zhang, Weiling Luan, Yuhang Yin, Fuqian Yang
ABSTRACT
Colloidal perovskite nanocrystals comprised of all inorganic cesium lead halide (CsPbX
Sigma-Aldrich
Cesium carbonate, 99.9% trace metals basis