Merck
  • DNA N6-methyladenine is dynamically regulated in the mouse brain following environmental stress.
DNA N6-methyladenine is dynamically regulated in the mouse brain following environmental stress.
Nature communications (2017-10-27)
Bing Yao, Ying Cheng, Zhiqin Wang, Yujing Li, Li Chen, Luoxiu Huang, Wenxin Zhang, Dahua Chen, Hao Wu, Beisha Tang, Peng Jin
ABSTRACT
Chemical modifications on DNA molecules, such as 5-methylcytosine and 5-hydroxymethylcytosine, play important roles in the mammalian brain. A novel DNA adenine modification, N(6)-methyladenine (6mA), has recently been found in mammalian cells. However, the presence and function(s) of 6mA in the mammalian brain remain unclear. Here we demonstrate 6mA dynamics in the mouse brain in response to environmental stress. We find that overall 6mA levels are significantly elevated upon stress. Genome-wide 6mA and transcriptome profiling reveal an inverse association between 6mA dynamic changes and a set of upregulated neuronal genes or downregulated LINE transposon expression. Genes bearing stress-induced 6mA changes significantly overlap with loci associated with neuropsychiatric disorders. These results suggest an epigenetic role for 6mA in the mammalian brain as well as its potential involvement in neuropsychiatric disorders.
Sigma-Aldrich
Phenol:Chloroform:Isoamyl Alcohol 25:24:1, Saturated with 10mM Tris, pH 8.0, 1mM EDTA, for molecular biology