Merck
Dinuclear nitrido-bridged ruthenium complexes bearing diimine ligands.
Dalton transactions (Cambridge, England : 2003) (2017-10-11)
Julie Urgiles, Sarah R Nathan, Samantha N MacMillan, Justin J Wilson
ABSTRACT
Reactions of K
Sigma-Aldrich
4-4′-Dimethoxy-2-2′-bipyridine, 97%