Merck
  • Fluorescent RNA cytosine analogue - an internal probe for detailed structure and dynamics investigations.
Fluorescent RNA cytosine analogue - an internal probe for detailed structure and dynamics investigations.
Scientific reports (2017-05-26)
Anders Foller Füchtbauer, Søren Preus, Karl Börjesson, Scott A McPhee, David M J Lilley, L Marcus Wilhelmsson
ABSTRACT
The bright fluorescent cytosine analogue tC
Sigma-Aldrich
Triethylamine trihydrofluoride, 97%