Merck
  • Doppler Parameters Derived from Transthoracic Echocardiography Accurately Detect Bioprosthetic Mitral Valve Dysfunction.
Doppler Parameters Derived from Transthoracic Echocardiography Accurately Detect Bioprosthetic Mitral Valve Dysfunction.
Journal of the American Society of Echocardiography : official publication of the American Society of Echocardiography (2017-08-22)
Ryan J Spencer, Ken G Gin, Michael Y C Tsang, Teresa S M Tsang, Parvathy Nair, Pui K Lee, John Jue
ABSTRACT
Detecting bioprosthetic mitral valve dysfunction on transthoracic echocardiography can be challenging because of acoustic shadowing of regurgitant jets and a wide normal range of transvalvular gradients. Several studies in mechanical mitral valves have demonstrated the utility of the transthoracically derived parameters E (peak early mitral inflow velocity), pressure half-time, and the ratio of mitral inflow velocity-time integral (VTI A total of 135 clinically stable patients with bioprosthetic mitral valves who had undergone both transthoracic and transesophageal echocardiography within a 6-month period were retrospectively identified from the past 11 years of the echocardiography database. Transthoracic findings were labeled as normal (n = 81), regurgitant (n = 44), or stenotic (n = 10) according to the patient's transesophageal echocardiographic findings. Univariate and multivariate analyses were performed to identify Doppler parameters that detected dysfunction; then receiver operating characteristic curves were created to establish appropriate normal cutoff levels. The VTI This study demonstrates that Doppler parameters derived from transthoracic echocardiography can accurately detect bioprosthetic mitral valve dysfunction. These parameters, particularly a VTI
Sigma-Aldrich
Pyrrolidone hydrotribromide, 97%