Merck
  • MGr1-Antigen/37 kDa laminin receptor precursor promotes cellular prion protein induced multi-drug-resistance of gastric cancer.
MGr1-Antigen/37 kDa laminin receptor precursor promotes cellular prion protein induced multi-drug-resistance of gastric cancer.
Oncotarget (2017-10-27)
Guanhong Luo, Weijie Wang, Qiong Wu, Yuanyuan Lu, Tao Su, Nan Gu, Kai Li, Jingbo Wang, Rui Du, Xiaodi Zhao, Xiaohua Li, Rui Fan, Hongbo Zhang, Yongzhan Nie, Xinmin Zhou, Yongquan Shi, Jie Liang, Xin Wang, Daiming Fan
ABSTRACT
Cellular prion protein (PrPC), the infective agent of transmissible spongiform encephalopathies, is thought to be related to several cellular physiological and physiopathological processes. We have previously reported that PrPC participates in multi-drug-resistance of gastric cancer. As the salient ligand molecule of PrP for participating in internalization and propagation of the scrapie form of prion protein (PrPSc), 37 kDa laminin receptor precursor protein (37LRP) shared the same gene coding sequence of MGr1-Ag, another protein previously found to be involved in multi-drug-resistance of gastric cancer in our lab. In the present study, we explored whether MGr1-Ag/37LRP contributed to PrPC mediated multi-drug-resistance in gastric cancer. Immunohistochemical staining showed similar expression patterns of MGr1-Ag/37LRP and PrPC in gastric cancer tissue serial sections. Western blot and immunohistochemistry also demonstrated correlative expression of MGr1-Ag/37LRP and PrPC in gastric cancer cell lines. Interaction between MGr1-Ag/37LRP and PrPC in gastric cancer cell lines and gastric cancer tissues were verified by immunofluorescence and co-immunoprecipitation. Furthermore, knockdown of MGr1-Ag/37LRP significantly attenuated PrPC induced multi-drug-resistance by sensitizing drug-induced apoptosis through inhibition of AKT activation. In conclusion, MGr1-Ag/37LRP may interact with PrPC and promote the PrPC induced multi-drug-resistance in gastric cancer through PI3K/AKT pathway. The current study elucidates the mechanism of how PrPC triggers intracellular signaling cascade resulting in multi-drug-resistance phenotype and provides a novel candidate molecular target against gastric cancer.
Sigma-Aldrich
MISSION® esiRNA, targeting human RPSA
Sigma-Aldrich
Anti-Prion Protein Antibody, a.a. 109-112, clone 3F4, clone 3F4, Chemicon®, from mouse