Merck
  • RNAi targeting Nogo Receptor enhanced survival and proliferation of murine retinal ganglion cells during N-methyl-D-aspartate-induced optic nerve crush.
RNAi targeting Nogo Receptor enhanced survival and proliferation of murine retinal ganglion cells during N-methyl-D-aspartate-induced optic nerve crush.
Oncotarget (2017-10-17)
Kun Zeng, Bo Zhong, Xiao-Li Shen, Min Fang, Bao-Tao Lin, Da-Hui Ma
ABSTRACT
We investigated the effects of lentivirus-mediated RNAi targeting of Nogo Receptor (
Sigma-Aldrich
Mechlorethamine hydrochloride, 98%
Sigma-Aldrich
N-Methyl-D-aspartic acid, ≥98% (TLC), solid