Merck
  • CXCL1 and CXCL2 Regulate NLRP3 Inflammasome Activation via G-Protein-Coupled Receptor CXCR2.
CXCL1 and CXCL2 Regulate NLRP3 Inflammasome Activation via G-Protein-Coupled Receptor CXCR2.
Journal of immunology (Baltimore, Md. : 1950) (2017-07-26)
Monoranjan Boro, Kithiganahalli Narayanaswamy Balaji
ABSTRACT
Inflammation is an extensively concerted process that confers protection to the host encountering immune insult. The major inflammatory mediators include IL-1 family members, such as IL-1β, and the functional activation of such molecules is arbitrated by their regulated cleavage brought about by components of a multiprotein complex called inflammasome. In this context, NLR family pyrin domain containing 3 (NLRP3) inflammasome activation often acts as a rate-limiting step in regulating critical cell-fate decisions in various inflammatory scenarios. In this study, we identify the G-protein-coupled receptor CXCR2 (recognizing chemokines CXCL1 and CXCL2) as another arm feeding into the regulated activation of NLRP3 inflammasome in macrophages. We demonstrate that in vivo blocking of CXCL1 and CXCL2 can significantly reduce the Mycobacterium tuberculosis-induced bioactive IL-1β production. Further, CXCL1 could amplify the inflammasome activation in in vivo mouse models of carrageenan-induced inflammation in footpads and air pouches. The mechanistic insights revealed CXCR2-driven protein kinase C μ-dependent integrin-linked kinase to be essential for CXCL1-mediated activation of NLRP3 inflammasome. Blocking the activity of integrin-linked kinase or protein kinase C μ either by small interfering RNA-mediated knockdown or pharmacological inhibitor compromised inflammasome activation and subsequent production of bioactive IL-1β. Taken together, our study demonstrates CXCR2-driven activation of NLRP3 inflammasome in macrophages and indicates a potential host-directed therapeutic target to limit the damaging inflammation associated with overt production of proinflammatory IL-1β.
Sigma-Aldrich
Anti-β-Actin−Peroxidase antibody, Mouse monoclonal, clone AC-15, purified from hybridoma cell culture
Sigma-Aldrich
CP-456773 sodium salt, ≥98% (HPLC)
Sigma-Aldrich
Anti-phospho-ILK (Ser246) Antibody, from rabbit, purified by affinity chromatography