Merck
Chloroquine inhibits Ca
Cell & bioscience (2017-05-27)
Yi-Fan Wu, Ping Zhao, Xi Luo, Jin-Chao Xu, Lu Xue, Qi Zhou, Mingrui Xiong, Jinhua Shen, Yong-Bo Peng, Meng-Fei Yu, Weiwei Chen, Liqun Ma, Qing-Hua Liu
ABSTRACT
Chloroquine, a bitter tastant, inhibits Ca In this study, thapsigargin (TG), but not caffeine, induced sustained intracellular Ca These results indicate that chloroquine inhibits Ca
Sigma-Aldrich
Pyr3, ≥98% (HPLC)
Sigma-Aldrich
Thapsigargin, ≥98% (HPLC), solid film