Merck
  • Pore geometry effect on the synthesis of silica supported perovskite oxides.
Pore geometry effect on the synthesis of silica supported perovskite oxides.
Journal of colloid and interface science (2017-06-06)
Jonas Scholz, Martin Etter, Diana Haas, Andreas Meyer, Andreas Kornowski, Uta Sazama, Simone Mascotto
ABSTRACT
The formation of perovskite oxide nanoparticles supported on ordered mesoporous silica with different pore geometry is here presented. Systematic study was performed varying both pore shape (gyroidal, cylindrical, spherical) and size (7.5, 12, 17nm) of the hosts. LaFeO
Sigma-Aldrich
Praseodymium(III) nitrate hexahydrate, 99.9% trace metals basis