- Pekinenin E Inhibits the Growth of Hepatocellular Carcinoma by Promoting Endoplasmic Reticulum Stress Mediated Cell Death.
Pekinenin E Inhibits the Growth of Hepatocellular Carcinoma by Promoting Endoplasmic Reticulum Stress Mediated Cell Death.
Frontiers in pharmacology (2017-07-15)
Lu Fan, Qingling Xiao, Yanyan Chen, Gang Chen, Jinao Duan, Weiwei Tao
PMID28706487
ABSTRACT
Hepatocellular carcinoma (HCC) is a malignant primary liver cancer with poor prognosis. In the present study, we report that pekinenin E (PE), a casbane diterpenoid derived from the roots of
MATERIALS