Merck
  • Identification of the Beer Component Hordenine as Food-Derived Dopamine D2 Receptor Agonist by Virtual Screening a 3D Compound Database.
Identification of the Beer Component Hordenine as Food-Derived Dopamine D2 Receptor Agonist by Virtual Screening a 3D Compound Database.
Scientific reports (2017-03-11)
Thomas Sommer, Harald Hübner, Ahmed El Kerdawy, Peter Gmeiner, Monika Pischetsrieder, Timothy Clark
ABSTRACT
The dopamine D2 receptor (D2R) is involved in food reward and compulsive food intake. The present study developed a virtual screening (VS) method to identify food components, which may modulate D2R signalling. In contrast to their common applications in drug discovery, VS methods are rarely applied for the discovery of bioactive food compounds. Here, databases were created that exclusively contain substances occurring in food and natural sources (about 13,000 different compounds in total) as the basis for combined pharmacophore searching, hit-list clustering and molecular docking into D2R homology models. From 17 compounds finally tested in radioligand assays to determine their binding affinities, seven were classified as hits (hit rate = 41%). Functional properties of the five most active compounds were further examined in β-arrestin recruitment and cAMP inhibition experiments. D2R-promoted G-protein activation was observed for hordenine, a constituent of barley and beer, with approximately identical ligand efficacy as dopamine (76%) and a K
Product Description
Supelco
Halofuginone hydrobromide, VETRANAL®, analytical standard
Supelco
Fenpropimorph, PESTANAL®, analytical standard
Supelco
Robenidine hydrochloride, VETRANAL®, analytical standard