Merck
  • Pairwise hydrogen addition in the selective semihydrogenation of alkynes on silica-supported Cu catalysts.
Pairwise hydrogen addition in the selective semihydrogenation of alkynes on silica-supported Cu catalysts.
Chemical science (2017-04-30)
Oleg G Salnikov, Hsueh-Ju Liu, Alexey Fedorov, Dudari B Burueva, Kirill V Kovtunov, Christophe Copéret, Igor V Koptyug
ABSTRACT
Mechanistic insight into the semihydrogenation of 1-butyne and 2-butyne on Cu nanoparticles supported on partially dehydroxylated silica (Cu/SiO
Sigma-Aldrich
2-Butyne, 99%