Merck
  • Immunohistochemical analysis of microsomal glutathione S-transferase 1 and clusterin expression in lens epithelial cells of patients with pseudoexfoliation syndrome.
Immunohistochemical analysis of microsomal glutathione S-transferase 1 and clusterin expression in lens epithelial cells of patients with pseudoexfoliation syndrome.
Experimental and therapeutic medicine (2017-04-30)
Joanna Stafiej, Marta Hałas-Wiśniewska, Magdalena Izdebska, Maciej Gagat, Dariusz Grzanka, Alina Grzanka, Grażyna Malukiewicz
ABSTRACT
Pseudoexfoliation syndrome (PEX) is an age-associated, sight disorder affecting elastic fibers in the eye and visceral organs but its exact etiology remains unknown. The purpose of the current study was to determine the morphology and ultrastructure of lens epithelial cells (LECs), and to use immunohistochemistry to examine localization of microsomal glutathione S-transferase 1 (MGST1) and clusterin. Anterior lens capsules were obtained from 24 patients (13 PEX and 11 controls) who underwent phacoemulsification. Immunohistochemistry was performed, using antibodies against MGST1 and clusterin, to determine their expression. The morphology and ultrastructure of LECs were evaluated by light and transmission electron microscopy, respectively. The PEX LECs were characterized by significantly lower MGST1 (P=0.0001) and clusterin expression (P=0.0005) compared with the control group patients. PEX LECs were also observed to have significantly increased thickness compared with the control group patients (P=0.0002). The current findings suggest that low MGST1 and clusterin expression levels may be an early clinical indicator of PEX, and that oxidative stress may serve an important role, but that the specific etiology of this disease has yet to be revealed.
Sigma-Aldrich
Anti-MGST1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
D.E.R. 332, used as embedding medium
Sigma-Aldrich
Anti-Clusterin antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution