Merck
  • A nitrate-selective electrode based on bis(triphenylphosphine)iminium salts.
A nitrate-selective electrode based on bis(triphenylphosphine)iminium salts.
Talanta (1989-09-01)
G Werner, I Kolowos, J Senkýr
ABSTRACT
A nitrate-sensitive electrode based on the ionic associates between bis(triphenylphosphine)iminium and nitrate ions in nitrobenzene solution is proposed. The electrode can be used for determinations of 10(-5) 0.1M nitrate in aqueous solutions.