Merck
  • Discrimination of red and white rice bran from Indonesia using HPLC fingerprint analysis combined with chemometrics.
Discrimination of red and white rice bran from Indonesia using HPLC fingerprint analysis combined with chemometrics.
Food chemistry (2016-12-17)
Aryani Sabir, Mohamad Rafi, Latifah K Darusman
ABSTRACT
HPLC fingerprint analysis combined with chemometrics was developed to discriminate between the red and the white rice bran grown in Indonesia. The major component in rice bran is γ-oryzanol which consisted of 4 main compounds, namely cycloartenol ferulate, cyclobranol ferulate, campesterol ferulate and β-sitosterol ferulate. Separation of these four compounds along with other compounds was performed using C18 and methanol-acetonitrile with gradient elution system. By using these intensity variations, principal component and discriminant analysis were performed to discriminate the two samples. Discriminant analysis was successfully discriminated the red from the white rice bran with predictive ability of the model showed a satisfactory classification for the test samples. The results of this study indicated that the developed method was suitable as quality control method for rice bran in terms of identification and discrimination of the red and the white rice bran.
Sigma-Aldrich
Campesterol, ~65%
Sigma-Aldrich
Cycloartenol, ≥90% (GC)