Merck
  • Functional Properties of Novel Epigallocatechin Gallate Glucosides Synthesized by Using Dextransucrase from Leuconostoc mesenteroides B-1299CB4.
Functional Properties of Novel Epigallocatechin Gallate Glucosides Synthesized by Using Dextransucrase from Leuconostoc mesenteroides B-1299CB4.
Journal of agricultural and food chemistry (2016-12-10)
Jiyoun Kim, Thi Thanh Hanh Nguyen, Nahyun M Kim, Young-Hwan Moon, Jung-Min Ha, Namhyeon Park, Dong-Gu Lee, Kyeong-Hwan Hwang, Jun-Seong Park, Doman Kim
ABSTRACT
Epigallocatechin gallate (EGCG) is the most abundant catechin found in the leaves of green tea, Camellia sinensis. In this study, novel epigallocatechin gallate-glucocides (EGCG-Gs) were synthesized by using dextransucrase from Leuconostoc mesenteroides B-1299CB4. Response surface methodology was adopted to optimize the conversion of EGCG to EGCG-Gs, resulting in a 91.43% conversion rate of EGCG. Each EGCG-G was purified using a C18 column. Of nine EGCG-Gs identified by nuclear magnetic resonance analysis, five EGCG-Gs (2 and 4-7) were novel compounds with yields of 2.2-22.6%. The water solubility of the five novel compounds ranged from 229.7 to 1878.5 mM. The 5'-OH group of EGCG-Gs expressed higher antioxidant activities than the 4'-OH group of EGCG-Gs. Furthermore, glucosylation at 7-OH group of EGCG-Gs was found to be responsible for maintaining tyrosinase inhibitory activity and increasing browning-resistant activities.