Merck
Brief Dark Exposure Reduces Tonic Inhibition in Visual Cortex.
The Journal of neuroscience : the official journal of the Society for Neuroscience (2015-12-04)
Shiyong Huang, Kristen Hokenson, Sabita Bandyopadhyay, Shelley J Russek, Alfredo Kirkwood
ABSTRACT
Tonic inhibition mediated by extrasynaptic GABA(A) receptors (GABARs) sensing ambient levels of GABA can profoundly alter the membrane input resistance to affect cellular excitability. Therefore, regulation of tonic inhibition is an attractive mechanism to control the levels of cortical firing. In cortical pyramidal cells, tonic inhibition is regulated by age and several neurotransmitters and is affected by stroke and epilepsy. However, the possible role of sensory experience has not been examined. Here, we report that a brief 2-day exposure to dark reduces by 1/3 the inhibitory tonic conductance recorded in layer II/III pyramidal cells of the mouse juvenile (postnatal day 12-27) visual cortex. In these cells, tonic inhibition is carried primarily by GABARs containing the δ subunit. Consistently, the dark exposure reduction in conductance was associated with a reduction in δ subunit levels, which were not affected in control frontal cortex. We propose that a deprivation-induced reduction in tonic inhibition might serve a homeostatic function by increasing the firing levels of cells in deprived cortical circuits.
Sigma-Aldrich
Anti-GABAA Receptor α1 Antibody, Upstate®, from rabbit
Sigma-Aldrich
Monoclonal Anti-β-Actin antibody produced in mouse, clone AC-15, ascites fluid