Merck
  • Boronate affinity-based surface molecularly imprinted polymers using glucose as fragment template for excellent recognition of glucosides.
Boronate affinity-based surface molecularly imprinted polymers using glucose as fragment template for excellent recognition of glucosides.
Journal of chromatography. A (2016-11-09)
Mijun Peng, Haiyan Xiang, Xin Hu, Shuyun Shi, Xiaoqing Chen
ABSTRACT
Rapid and efficient extraction of bioactive glycosides from complex natural origins poses a difficult challenge, and then is often inherent bottleneck for their highly utilization. Herein, we propose a strategy to fabricate boronate affinity based surface molecularly imprinted polymers (MIPs) for excellent recognition of glucosides. d-glucose was used as fragment template. Boronic acid, dynamic covalent binding with d-glucose under different pH conditions, was selected as functional monomer to improve specificity. Fe
Sigma-Aldrich
3-Formylphenylboronic acid, ≥95%