Merck
  • A new class of ammonium ylid for [2,3]-sigmatropic rearrangement reactions: ene-endo-spiro ylids.
A new class of ammonium ylid for [2,3]-sigmatropic rearrangement reactions: ene-endo-spiro ylids.
Organic letters (2005-05-07)
Edward Roberts, Julien P Sançon, J B Sweeney
ABSTRACT
The first examples of sigmatropic rearrangements of ene-endo-spirocyclic, tetrahydropyridine-derived ammonium ylids are reported. Thus, spiro[6.7]-ylids rearrange primarily by a [2,3]-pathway, whereas the analogous [6.6]-ylids rearrange by [1,2]- and [2,3]-mechanisms in roughly equal proportions. This method serves as a rapid entry to the core of a range of alkaloids bearing a pyrrolo[1,2-a]azepine or octahydroindolizidine nucleus.
Sigma-Aldrich
1,2,3,6-Tetrahydropyridine, 97%