Merck
  • Enol tosylates as viable partners in Pd-catalyzed cross-coupling reactions.
Enol tosylates as viable partners in Pd-catalyzed cross-coupling reactions.
The Journal of organic chemistry (2005-11-19)
Dietrich Steinhuebel, Jenny M Baxter, Michael Palucki, Ian W Davies
ABSTRACT
[reaction: see text] Herein we demonstrate functionalized enol tosylates to be robust substrates that undergo Suzuki-Miyaura, Sonogashira, and Stille cross-coupling reactions to provide stereodefined trisubstituted unsaturated esters.
Sigma-Aldrich
Methylboronic acid, 97%