Merck
Allosteric inhibitors of Bcr-abl-dependent cell proliferation.
Nature chemical biology (2006-01-18)
Francisco J Adrián, Qiang Ding, Taebo Sim, Anastasia Velentza, Christine Sloan, Yi Liu, Guobao Zhang, Wooyoung Hur, Sheng Ding, Paul Manley, Jürgen Mestan, Doriano Fabbro, Nathanael S Gray
ABSTRACT
Chronic myelogenous leukemia (CML) is a myeloproliferative disorder characterized at the molecular level by the expression of Bcr-abl, a 210-kDa fusion protein with deregulated tyrosine kinase activity. Encouraged by the clinical validation of Bcr-abl as the target for the treatment of CML by imatinib, we sought to identify pharmacological agents that could target this kinase by a distinct mechanism. We report the discovery of a new class of Bcr-abl inhibitors using an unbiased differential cytotoxicity screen of a combinatorial kinase-directed heterocycle library. Compounds in this class (exemplified by GNF-2) show exclusive antiproliferative activity toward Bcr-abl-transformed cells, with potencies similar to imatinib, while showing no inhibition of the kinase activity of full-length or catalytic domain of c-abl. We propose that this new class of compounds inhibits Bcr-abl kinase activity through an allosteric non-ATP competitive mechanism.
Sigma-Aldrich
Adenosine 5′-triphosphate (ATP) disodium salt hydrate, vial of ~1 mg ATP
Sigma-Aldrich
GNF-2, ≥98% (HPLC), solid