Merck
  • Natural Genetic Variation Influences Protein Abundances in C. elegans Developmental Signalling Pathways.
Natural Genetic Variation Influences Protein Abundances in C. elegans Developmental Signalling Pathways.
PloS one (2016-03-18)
Kapil Dev Singh, Bernd Roschitzki, L Basten Snoek, Jonas Grossmann, Xue Zheng, Mark Elvin, Polina Kamkina, Sabine P Schrimpf, Gino B Poulin, Jan E Kammenga, Michael O Hengartner
ABSTRACT
Complex traits, including common disease-related traits, are affected by many different genes that function in multiple pathways and networks. The apoptosis, MAPK, Notch, and Wnt signalling pathways play important roles in development and disease progression. At the moment we have a poor understanding of how allelic variation affects gene expression in these pathways at the level of translation. Here we report the effect of natural genetic variation on transcript and protein abundance involved in developmental signalling pathways in Caenorhabditis elegans. We used selected reaction monitoring to analyse proteins from the abovementioned four pathways in a set of recombinant inbred lines (RILs) generated from the wild-type strains N2 (Bristol) and CB4856 (Hawaii) to enable quantitative trait locus (QTL) mapping. About half of the cases from the 44 genes tested showed a statistically significant change in protein abundance between various strains, most of these were however very weak (below 1.3-fold change). We detected a distant QTL on the left arm of chromosome II that affected protein abundance of the phosphatidylserine receptor protein PSR-1, and two separate QTLs that influenced embryonic and ionizing radiation-induced apoptosis on chromosome IV. Our results demonstrate that natural variation in C. elegans is sufficient to cause significant changes in signalling pathways both at the gene expression (transcript and protein abundance) and phenotypic levels.
Product Description
Supelco
Bradford Reagent, for 0.1-1.4 mg/ml protein
Sigma-Aldrich
Glass beads, acid-washed, 212-300 μm (50-70 U.S. sieve)