Merck
  • Chrysamine G and its derivative reduce amyloid beta-induced neurotoxicity in mice.
Chrysamine G and its derivative reduce amyloid beta-induced neurotoxicity in mice.
Neuroscience letters (2002-10-29)
Kazuhiro Ishii, William E Klunk, Shigeki Arawaka, Manik L Debnath, Yoshiko Furiya, Naruhiko Sahara, Shin'ichi Shoji, Akira Tamaoka, Jay W Pettegrew, Hiroshi Mori
ABSTRACT
The neurotoxicity of amyloid beta (Abeta) is widely believed to play a seminal role in neurodegeneration in Alzheimer's disease. We examined the effect of Chrysamine G (CG) on such neurotoxicity using the specific measurement of surviving neurons. CG was found to reduce the neurodegeneration induced by both the active short fragment of Abeta(25-35) and full-sized Abeta(1-40). In this study, we synthesized a new chemical compound from a monovalent structure of CG (hCG), with a lower affinity for Abeta, and compared its activity with that of CG. Both CG and hCG were found to be equally efficacious in reducing Abeta-induced neuronal death at a concentration of 0.1-1 microM, indicating that the mechanism of action for CG was not due to its chelating activity, but rather due to its anti-oxidant activity.
Sigma-Aldrich
Chrysamine G, ≥95% (HPLC), solid