Merck
  • HIV-1 protease inhibitors based on hydroxyethylene dipeptide isosteres: an investigation into the role of the P1' side chain on structure-activity.
HIV-1 protease inhibitors based on hydroxyethylene dipeptide isosteres: an investigation into the role of the P1' side chain on structure-activity.
Journal of medicinal chemistry (1992-05-15)
S D Young, L S Payne, W J Thompson, N Gaffin, T A Lyle, S F Britcher, S L Graham, T H Schultz, A A Deana, P L Darke
ABSTRACT
A systematic investigation was undertaken to determine the role of the P1' sidechain in a series of hydroxyethylene isostere based inhibitors of HIV-1 protease. Substitution and homologation of the benzyl P1' side chain of the Phe-Phe isostere based pseudo peptides 1 (L-682,679) and 2 (L-685,434) with various heteroalkyl groups leads to a series of extremely potent inhibitors of the enzyme. Several examples of the most potent inhibitors were very effective in an ex vivo cell based viral spread assay using human H9 T-lymphocytes and the IIIb isolate of HIV-1. Compound 19 is 120 times more potent than 1 and 16 times more potent than 2 in inhibiting the spread of infection in this assay.
Sigma-Aldrich
(1R,2S)-(+)-cis-1-Amino-2-indanol, 99%