Merck
  • [Helicobacter pylori erradication and its relation to antibiotic resistance and CYP2C19 status].
[Helicobacter pylori erradication and its relation to antibiotic resistance and CYP2C19 status].
Revista espanola de enfermedades digestivas : organo oficial de la Sociedad Espanola de Patologia Digestiva (2007-04-10)
E Garza González, E Giasi González, M A Martínez Vázquez, G I Pérez Pérez, G M González, H J Maldonado Garza, F J Bosques Padilla
ABSTRACT
to assess the efficacy of rabeprazole (RPZ), amoxicillin (Am), and clarithromycin (Cla) (7 vs. 14 days) in the eradication of H. pylori, and to determine the effect of strain-specific antibiotic resistance and host CYP2C19 status. first, we determined the CYP2C19 status of 100 healthy subjects to establish a sample size for the clinical trial. Then, 59 H. pylori-infected patients were randomized to receive RPZ (20 mg daily) plus Cla (500 mg b.d.) and Am (1,000 mg b.d.) for 7 vs. 14 days. The MIC for Am and Cla were determined using the agar dilution method. The CYP2C19 genotype was determined by the PCR-RFLP method. In the per-protocol analysis (PP) eradication rates were 89.7 and 72% for the 7- and 14-day groups (p = 0.159). In the intention to-treat analysis (ITT) eradication rates were 86.7 and 62.1% in the 7- and 14-day groups, respectively (p = 0.06). None of the strains was resistant to Am, and 4 strains were resistant to Cla: 3 (11.1%) in the 14-day group and 1 (4%) in the 7-day group. Neither strain-specific antibiotic resistance nor host CYP2C19 status influenced eradication rates. both 7- and 14-day therapies were effective for H. pylori eradication. Strain resistance and CYP2C19 status do not seem to influence eradication rates in the studied population.