Merck
An improved efficient synthesis of the antibacterial agent torezolid.
Chemical & pharmaceutical bulletin (2014-12-11)
Gang Li, Bao-Kun Yuan, Wu Tang, Hong-Yi Zhao, Zi-Yun Lin, Hai-Hong Huang
ABSTRACT
An improved and efficient method for the preparation of torezolid based on Suzuki cross-coupling reaction as the key step was developed on a gram scale in five steps. The total yield was 44% and the optical purity of torezolid by the improved method was above 99%.
Sigma-Aldrich
4-Bromo-3-fluoroaniline, 97%