Merck
  • A facile method for controlling the reaction equilibrium of sphingolipid ceramide N-deacylase for lyso-glycosphingolipid production.
A facile method for controlling the reaction equilibrium of sphingolipid ceramide N-deacylase for lyso-glycosphingolipid production.
Journal of lipid research (2015-07-02)
Feng-Tao Huang, Yun-Bin Han, Yan Feng, Guang-Yu Yang
ABSTRACT
Lyso-glycosphingolipids (lyso-GSLs), the N-deacylated forms of glycosphingolipids (GSLs), are important synthetic intermediates for the preparation of GSL analogs. Although lyso-GSLs can be produced by hydrolyzing natural GSLs using sphingolipid ceramide N-deacylase (SCDase), the yield for this reaction is usually low because SCDase also catalyzes the reverse reaction, ultimately establishing an equilibrium between hydrolysis and synthesis. In the present study, we developed an efficient method for controlling the reaction equilibrium by introducing divalent metal cation and detergent in the enzymatic reaction system. In the presence of both Ca(2+) and taurodeoxycholate hydrate, the generated fatty acids were precipitated by the formation of insoluble stearate salts and pushing the reaction equilibrium toward hydrolysis. The yield of GM1 hydrolysis can be achieved as high as 96%, with an improvement up to 45% compared with the nonoptimized condition. In preparative scale, 75 mg of lyso-GM1 was obtained from 100 mg of GM1 with a 90% yield, which is the highest reported yield to date. The method can also be used for the efficient hydrolysis of a variety of GSLs and sphingomyelin. Thus, this method should serve as a facile, easily scalable, and general tool for lyso-GSL production to facilitate further GSL research.
Sigma-Aldrich
Sphingosylphosphorylcholine, ≥98%, powder