Merck
  • Surface-enhanced Raman scattering behaviour of 4-mercaptophenyl boronic acid on assembled silver nanoparticles.
Surface-enhanced Raman scattering behaviour of 4-mercaptophenyl boronic acid on assembled silver nanoparticles.
Physical chemistry chemical physics : PCCP (2015-06-18)
Shuangshuang Li, Qun Zhou, Wenya Chu, Wei Zhao, Junwei Zheng
ABSTRACT
Molecular recognition based on specific intermolecular interactions is essential for the design of sensors with high selectivity. Herein, we report the surface-enhanced Raman scattering (SERS) behaviour of 4-mercaptophenyl boronic acid (MPBA) on self-assembled silver nanoparticles and its interaction with d-glucose. It is demonstrated that the orientation and existing form of the MPBA strongly depend on the pH value of the media. The surface-immobilized MPBA can be reversibly associated with OH(-) in solution, along with a molecular orientation alteration. A self-condensation reaction among the OH(-)-associated MPBA molecules results in irreversible conversion of OH(-)-associated MPBA to anhydride, which may hinder the interaction between d-glucose and the B-moiety of MPBA. However, the self-condensation reaction can be diminished under optimized conditions. By taking advantage of the difference in the kinetics of dissociation of the OH(-)-associated MPBA and d-glucose-associated MPBA in acidic media, a proper scheme of the SERS detection of d-glucose is proposed to illuminate the spectral interference of OH(-)-associated MPBA, which exhibits SERS features similar to those of d-glucose-associated MPBA species. Based on those strategies, the SERS detection of d-glucose can be achieved in the physiologically-relevant concentration range.
Sigma-Aldrich
4-Mercaptophenylboronic acid, 90%