Merck
Variants of interleukin-16 associated with gastric cancer risk.
Asian Pacific journal of cancer prevention : APJCP (2013-11-02)
Tao Zhang, Hui Wang
ABSTRACT
We conducted a case-control matched study to investigate the role of IL-16 gene polymorphisms, rs4072111, rs1131445, rs4778889 and rs11556218, in the risk of gastric cancer in a Chinese population, also performing subgroup analysis by subsites. To test the hypothesis of involvement, we analyzed the four SNPs of IL16 in 347 cancer patients and 368 controls. Demographic data and other information were collected using a newly designed questionnaire. Genotyping of IL16 (rs4072111, rs1131445, rs4778889 and rs11556218) was performed in a 384-well plate format on the MassARRAY® platform. In our study, we found the gastric cancer patients were more likely to be male and have a family history of cancer (P < 0.05). We found the rs4778889 CC and rs11556218 GG genotype was significantly associated with 1.97 and 1.84-fold increased risk of non-cardia gastric cancer, while we did not find significant association between the four IL-16 SNPs and cardia gastric cancer. In conclusion, our study indicated that IL-16 rs4778889 CC and rs11556218 GG genotypes are associated with an increased risk of non-cardia gastric cancer in a Chinese population. Our results offer insights into the influence of IL-16 on development of gastric cancer.