Merck
  • Somatostatin receptor 5 is palmitoylated by the interacting ZDHHC5 palmitoyltransferase.
Somatostatin receptor 5 is palmitoylated by the interacting ZDHHC5 palmitoyltransferase.
FEBS letters (2011-08-09)
Tarja Kokkola, Claudia Kruse, Eva-Maria Roy-Pogodzik, Jenna Pekkinen, Carola Bauch, Hans-Hinrich Hönck, Hanjo Hennemann, Hans-Jürgen Kreienkamp
ABSTRACT
Many G-protein coupled receptors are palmitoylated in their C-terminal, intracellular regions. So far no enzymes responsible for this modification have been described. We identified an interaction of the membrane proximal helix 8 of somatostatin receptor 5 (SSTR5) with the N-terminal region of the putative palmitoyltransferase ZDHHC5 using the Ras recruitment interaction screening system. ZDHHC5 and SSTR5 are colocalized at the plasma membrane and can be efficiently coimmunoprecipitated from transfected cells. Coexpression of ZDHHC5 in HEK293 cells increased palmitoylation of SSTR5 whereas knock-down of endogenous ZDHHC5 by siRNAs decreased it. Our data identify the first palmitoyltransferase for a G-protein coupled receptor.