- Syntheses of deuterium labeled prenyldiphosphate and prenylcysteine analogues for in vivo mass spectrometric quantification.
Syntheses of deuterium labeled prenyldiphosphate and prenylcysteine analogues for in vivo mass spectrometric quantification.
Journal of labelled compounds & radiopharmaceuticals (2013-11-29)
Thangaiah Subramanian, Karunai Leela Subramanian, Manjula Sunkara, Fredrick O Onono, Andrew J Morris, H Peter Spielmann
PMID24285475
ABSTRACT
A Wittig reaction employing Li(CD3)2CP(C6H5)3 was used to prepare d6-farnesol and d6-geranylgeraniol. Reductive amination of aniline-2,3,4,5,6-d5 was used to prepare the unnatural isoprenoid analogues d5-anilinogeraniol and d5-anilinofarnesol. All of these deuterated isoprenols were elaborated into their diphosphate and cysteine thioether derivatives suitable for use as stable-isotope labeled standards for quantitative mass spectrometric analysis.