Merck
  • Toward selective drug development for the human 5-hydroxytryptamine 1E receptor: a comparison of 5-hydroxytryptamine 1E and 1F receptor structure-affinity relationships.
Toward selective drug development for the human 5-hydroxytryptamine 1E receptor: a comparison of 5-hydroxytryptamine 1E and 1F receptor structure-affinity relationships.
The Journal of pharmacology and experimental therapeutics (2011-03-23)
Michael T Klein, Małgorzata Dukat, Richard A Glennon, Milt Teitler
ABSTRACT
The 5-hydroxytryptamine (5-HT) 1E receptor is highly expressed in the human frontal cortex and hippocampus, and this distribution suggests the function of 5-HT(1E) receptors might be linked to memory. To test this hypothesis, behavioral experiments are needed. Because rats and mice lack a 5-HT(1E) receptor gene, knockout strategies cannot be used to elucidate this receptor's functions. Thus, selective pharmacological tools must be developed. The tryptamine-related agonist BRL54443 [5-hydroxy-3-(1-methylpiperidin-4-yl)-1H-indole] is one of the few agents that binds 5-HT(1E) receptors with high affinity and some selectively; unfortunately, it binds equally well to 5-HT(1F) receptors (K(i) ≈ 1 nM). The differences between tryptamine binding requirements of these two receptor populations have never been extensively explored; this must be done to guide the design of analogs with greater selectivity for 5-HT(1E) receptors versus 5-HT(1F) receptors. Previously, we determined the receptor binding affinities of a large series of tryptamine analogs at the 5-HT(1E) receptor; we now examine the affinities of this same series of compounds at 5-HT(1F) receptors. The affinities of these compounds at 5-HT(1E) and 5-HT(1F) receptors were found to be highly correlated (r = 0.81). All high-affinity compounds were full agonists at both receptor populations. We identified 5-N-butyryloxy-N,N-dimethyltryptamine as a novel 5-HT(1F) receptor agonist with >60-fold selectivity versus 5-HT(1E) receptors. There is significant overlap between 5-HT(1E) and 5-HT(1F) receptor orthosteric binding properties; thus, identification of 5-HT(1E)-selective orthosteric ligands will be difficult. The insights generated from this study will inform future drug development and molecular modeling studies for both 5-HT(1E) and 5-HT(1F) receptors.