Merck
Total synthesis of entecavir.
The Journal of organic chemistry (2013-05-18)
Javier Velasco, Xavier Ariza, Laura Badía, Martí Bartra, Ramon Berenguer, Jaume Farràs, Joan Gallardo, Jordi Garcia, Yolanda Gasanz
ABSTRACT
Entecavir (BMS-200475) was synthesized from 4-trimethylsilyl-3-butyn-2-one and acrolein. The key features of its preparation are: (i) a stereoselective boron-aldol reaction to afford the acyclic carbon skeleton of the methylenecylopentane moiety; (ii) its cyclization by a Cp2TiCl-catalyzed intramolecular radical addition of an epoxide to an alkyne; and (iii) the coupling with a purine derivative by a Mitsunobu reaction.
Sigma-Aldrich
4-(Trimethylsilyl)-3-butyn-2-one, 97%