Merck
  • Pivotal role for ROS activation of p38 MAPK in the control of differentiation and tumor-initiating capacity of glioma-initiating cells.
Pivotal role for ROS activation of p38 MAPK in the control of differentiation and tumor-initiating capacity of glioma-initiating cells.
Stem cell research (2013-11-05)
Atsushi Sato, Masashi Okada, Keita Shibuya, Eriko Watanabe, Shizuka Seino, Yoshitaka Narita, Soichiro Shibui, Takamasa Kayama, Chifumi Kitanaka
ABSTRACT
Reactive oxygen species (ROS) are involved in various aspects of cancer cell biology, yet their role in cancer stem cells (CSCs) has been poorly understood. In particular, it still remains unclear whether and how ROS control the self-renewal/differentiation process and the tumor-initiating capacity of CSCs. Here we show that ROS-mediated activation of p38 MAPK plays a pivotal role in the control of differentiation and tumor-initiating capacity of glioma-initiating cells (GICs) derived from human glioblastomas. Mechanistically, ROS triggered p38-dependent Bmi1 protein degradation and FoxO3 activation in GICs, which were shown to be responsible for the loss of their self-renewal capacity and differentiation, respectively. Thus, the results suggest that Bmi1 and FoxO3 govern distinct phases of transition from undifferentiated to fully differentiated cells. Furthermore, we also demonstrate in this study that oxidative stress deprives GICs of their tumor-initiating capacity through the activation of the ROS-p38 axis. As such, this is the first study to the best of our knowledge to delineate how ROS control self-renewal/differentiation and the tumor-initiating capacity of stem-like cancer cells. This study also suggests that targeting of the ROS-p38 axis could be a novel approach in the development of therapeutic strategies against gliomas, represented by glioblastoma.
Sigma-Aldrich
Cycloheximide, Biotechnology Performance Certified
Sigma-Aldrich
SL327, ≥98% (HPLC), Mixture of E & Z isomers, solid
Sigma-Aldrich
L-Buthionine-sulfoximine, ≥97% (TLC)
Sigma-Aldrich
Cycloheximide, from microbial, ≥94% (TLC)
Sigma-Aldrich
Cycloheximide, ≥95% (HPLC)
Supelco
Cycloheximide, PESTANAL®, analytical standard
Sigma-Aldrich
LY 294002, LY294002, CAS 154447-36-6, is a cell-permeable, potent, reversible, and specific inhibitor of PI 3-kinase ((IC₅₀ = 1.4 µM). Acts on the ATP-binding site.
Sigma-Aldrich
Anti-Bmi-1 Antibody, clone F6, clone F6, Upstate®, from mouse
Sigma-Aldrich
Anti-Neurofilament NF-H Antibody, tail domain (Phosphate dependent), clone RMO-24, ascites fluid, clone RMO-24, Chemicon®
Sigma-Aldrich
Anti-β-Actin antibody, Mouse monoclonal, clone AC-15, purified from hybridoma cell culture
Millipore
Cycloheximide solution, 0.1%, suitable for microbiology
Sigma-Aldrich
Cycloheximide solution, Ready-Made Solution, microbial, 100 mg/mL in DMSO, Suitable for cell culture
Sigma-Aldrich
N-Acetyl-L-cysteine, BioReagent, suitable for cell culture
Sigma-Aldrich
Z-Leu-Leu-Leu-al, ≥90% (HPLC)
Sigma-Aldrich
SB 203580, solid, ≥98% (HPLC)
Sigma-Aldrich
Anti-α-Tubulin Mouse mAb (DM1A), liquid, clone DM1A, Calbiochem®