Merck
  • Human brain sodium channels as one of the molecular target sites for the new intravenous anaesthetic propofol (2,6-diisopropylphenol).
Human brain sodium channels as one of the molecular target sites for the new intravenous anaesthetic propofol (2,6-diisopropylphenol).
European journal of pharmacology (1991-09-12)
C Frenkel, B W Urban
ABSTRACT
Single sodium channels from human brain cortex tissue were incorporated into voltage-clamped planar lipid bilayers in the presence of batrachotoxin and studied with various doses of the new anaesthetic compound propofol (2,6-diisopropylphenol). Propofol was found to depress two major sodium channel functions, leading to a reduction of the time-averaged fractional channel open-time (half-maximal blocking concentration, ED50, 20 microM; maximal block 28%) and an interaction with the steady-state activation. These effects occurred at clinically relevant serum concentrations, suggesting the human brain sodium channel as one of the molecular target sites of action for propofol.
Sigma-Aldrich
2,6-Diisopropylphenol, 97%