Merck
  • Enhanced durability of polymer electrolyte membrane fuel cells by functionalized 2D boron nitride nanoflakes.
Enhanced durability of polymer electrolyte membrane fuel cells by functionalized 2D boron nitride nanoflakes.
ACS applied materials & interfaces (2014-04-09)
Keun-Hwan Oh, Dongju Lee, Min-Ju Choo, Kwang Hyun Park, Seokwoo Jeon, Soon Hyung Hong, Jung-Ki Park, Jang Wook Choi
ABSTRACT
We report boron nitride nanoflakes (BNNFs), for the first time, as a nanofiller for polymer electrolyte membranes in fuel cells. Utilizing the intrinsic mechanical strength of two-dimensional (2D) BN, addition of BNNFs even at a marginal content (0.3 wt %) significantly improves mechanical stability of the most representative hydrocarbon-type (HC-type) polymer electrolyte membrane, namely sulfonated poly(ether ether ketone) (sPEEK), during substantial water uptake through repeated wet/dry cycles. For facile processing with BNNFs that frequently suffer from poor dispersion in most organic solvents, we non-covalently functionalized BNNFs with 1-pyrenesulfonic acid (PSA). Besides good dispersion, PSA supports efficient proton transport through its sulfonic functional groups. Compared to bare sPEEK, the composite membrane containing BNNF nanofiller exhibited far improved long-term durability originating from enhanced dimensional stability and diminished chronic edge failure. This study suggests that introduction of properly functionalized 2D BNNFs is an effective strategy in making various HC-type membranes sustainable without sacrificing their original adventurous properties in polymer electrolyte membrane fuel cells.
Sigma-Aldrich
1-Pyrenesulfonic acid hydrate