Merck
  • Graphene-wrapped MnO2 -graphene nanoribbons as anode materials for high-performance lithium ion batteries.
Graphene-wrapped MnO2 -graphene nanoribbons as anode materials for high-performance lithium ion batteries.
Advanced materials (Deerfield Beach, Fla.) (2013-09-03)
Lei Li, Abdul-Rahman O Raji, James M Tour
ABSTRACT
A facile and cost-effective approach for the fabrication of a hierarchical nanocomposite material of graphene-wrapped MnO2 -graphene nanoribbons (GMG) is developed. The resulting composite has a high specific capacity and an excellent cycling stability owing to the synergistic combination of the electrically conductive graphene, graphene nanoribbons, and MnO2 .