Merck
  • A rapid and general method for the asymmetric synthesis of 2-substituted pyrrolidines using tert-butanesulfinamide.
A rapid and general method for the asymmetric synthesis of 2-substituted pyrrolidines using tert-butanesulfinamide.
Organic & biomolecular chemistry (2005-05-27)
Kristin M Brinner, Jonathan A Ellman
ABSTRACT
A new method for the asymmetric synthesis of 2-substituted pyrrolidines in three steps from commercially available starting materials is described. Addition of the Grignard reagent prepared from 2-(2-bromoethyl)-1,3-dioxane to N-tert-butanesulfinyl aldimines proceeds in high yields and with good diastereoselectivities. The sulfinamide products are then cleanly converted into pyrrolidines in one step.