- Blood levels and distribution of talampicillin and bacampicillin in NZW rabbits.
Blood levels and distribution of talampicillin and bacampicillin in NZW rabbits.
The Tokai journal of experimental and clinical medicine (1986-05-01)
N Yamane, T Konnai, T Imoto, T Sekiguchi, S Tomita, Y Ohta, J Sasaki
PMID3564081
ABSTRACT
Parameters for blood concentrations of the peroral antibiotics talampicil in and bacampicillin, in NZW rabbits were determined using a two compartment model. On the basis of these pharmacokinetic parameters, a three compartment model was prepared for the concentrations of these drugs in the tongue, gingiva, submandibular gland, parotid gland, cervical lymph-node and mandibular bone. Simulation curves based on the parameters of tissue concentrations revealed visual conformity to the measured values.
Product Description
Talampicillin hydrochloride, European Pharmacopoeia (EP) Reference Standard