Merck
Managing an academic career.
The American journal of occupational therapy : official publication of the American Occupational Therapy Association (1986-02-01)
G N Masagatani, H K Grant
ABSTRACT
Current predicaments that academic educators deal with, that is, the conflicts between teaching and research responsibilities, the competition within universities for promotion and tenure, and the importance of viewing the university as a complex social-political system are addressed. Recommendations for improvements in the development and education of new faculty members and suggestions for evaluative criteria for the documentation of a faculty member's performance are offered.
Sigma-Aldrich
Ginseng root from Panax quinquefolium (American ginseng)