Merck
  • Effect of nortriptyline on symptoms of idiopathic gastroparesis: the NORIG randomized clinical trial.
Effect of nortriptyline on symptoms of idiopathic gastroparesis: the NORIG randomized clinical trial.
JAMA (2013-12-26)
Henry P Parkman, Mark L Van Natta, Thomas L Abell, Richard W McCallum, Irene Sarosiek, Linda Nguyen, William J Snape, Kenneth L Koch, William L Hasler, Gianrico Farrugia, Linda Lee, Aynur Unalp-Arida, James Tonascia, Frank Hamilton, Pankaj J Pasricha
ABSTRACT
Gastroparesis remains a challenging syndrome to manage, with few effective treatments and a lack of rigorously controlled trials. Tricyclic antidepressants are often used to treat refractory symptoms of nausea, vomiting, and abdominal pain. Evidence from well-designed studies for this use is lacking. To determine whether treatment with nortriptyline results in symptomatic improvement in patients with idiopathic gastroparesis. The NORIG (Nortriptyline for Idiopathic Gastroparesis) trial, a 15-week multicenter, parallel-group, placebo-controlled, double-masked, randomized clinical trial from the National Institute of Diabetes and Digestive and Kidney Diseases Gastroparesis Clinical Research Consortium (GpCRC), comparing nortriptyline with placebo for symptomatic relief in idiopathic gastroparesis. One hundred thirty patients with idiopathic gastroparesis were enrolled between March 2009 and June 2012 at 7 US academic medical centers. Patient follow-up was completed in October 2012. Inclusion criteria included delayed gastric emptying and moderate to severe symptom scores using the Gastroparesis Cardinal Symptom Index (GCSI). INTERVENTIONS Nortriptyline vs placebo. Study drug dose was increased at 3-week intervals (10, 25, 50, 75 mg) up to 75 mg at 12 weeks. The primary outcome measure of symptomatic improvement was a decrease from the patient's baseline GCSI score of at least 50% on 2 consecutive 3-week GCSI assessments during 15 weeks of treatment. The primary symptomatic improvement outcome did not differ between 65 patients randomized to nortriptyline vs 65 patients randomized to placebo: 15 (23% [95% CI, 14%-35%]) in the nortriptyline group vs 14 (21% [95% CI, 12%-34%]) in the placebo group (P = .86). Treatment was stopped more often in the nortriptyline group (19 [29% {95% CI, 19%-42%}]) than in the placebo group (6 [9%] {95% CI, 3%-19%}]) (P = .007), but numbers of adverse events were not different (27 [95% CI, 18-39] vs 28 [95% CI, 19-40]) (P = .89). Among patients with idiopathic gastroparesis, the use of nortriptyline compared with placebo for 15 weeks did not result in improvement in overall symptoms. These findings do not support the use of nortriptyline for idiopathic gastroparesis. clinicaltrials.gov Identifier: NCT00765895.
Sigma-Aldrich
Nortriptyline hydrochloride, ≥98% (HPLC), powder
Nortriptyline hydrochloride, European Pharmacopoeia (EP) Reference Standard
Supelco
Nortriptyline hydrochloride solution, 1.0 mg/mL in methanol (as free base), ampule of 1 mL, certified reference material, Cerilliant®