Merck
2-Chloro-6-fluoro-benzoic acid.
Acta crystallographica. Section E, Structure reports online (2011-07-15)
Richard Betz, Thomas Gerber
ABSTRACT
The title compound, C(7)H(4)ClFO(2), is a twofold halogenated derivative of benzoic acid. The C-C-C angles within the aromatic moiety cover a range 116.11 (14)-123.96 (15)°, with the maximum and the minimum value next to each other. In the crystal, O-H⋯O hydrogen bonds form carb-oxy-lic acid dimers, which are further connected by C-H⋯F contacts into undulating sheets perpendicular to the a axis.