Merck
  • Synthesis of dicationic ionic liquids and their application in the preparation of hierarchical zeolite Beta.
Synthesis of dicationic ionic liquids and their application in the preparation of hierarchical zeolite Beta.
Chemistry (Weinheim an der Bergstrasse, Germany) (2011-11-25)
Rajkumar Kore, Biswarup Satpati, Rajendra Srivastava
ABSTRACT
Piperidine- and imidazole-based dicatoinic ionic liquids have been developed for the synthesis of zeolite Beta. Hierarchical Beta has a larger surface area and pore volume than conventional Beta. Beta derived from a dicationic ionic liquid exhibited remarkably higher catalytic activity than the conventional Beta. Experimental evidence and DFT calculations suggest that only a suitable conformation of such dicationic ionic liquids is able to form zeolite Beta (see scheme).
Sigma-Aldrich
4,4′-Trimethylenebis(1-methylpiperidine), ≥98%