Merck
  • Palladium-catalyzed copolymerization of ethene with acrolein dimethyl acetal: catalyst action and deactivation.
Palladium-catalyzed copolymerization of ethene with acrolein dimethyl acetal: catalyst action and deactivation.
Journal of the American Chemical Society (2004-09-30)
Weidong Li, Xiaochun Zhang, Auke Meetsma, Bart Hessen
ABSTRACT
Acrolein dimethyl acetal (ADMA) can be copolymerized with ethene using a cationic alpha-diimine palladium chelate catalyst to yield branched copolymers. Catalyst deactivation occurs via methanol elimination to give an inert eta3-1-methoxyallyl palladium species. This process can be retarded by the addition of an aliquot of methanol to the reaction mixture, increasing overall catalyst productivity.
Sigma-Aldrich
Acrolein dimethyl acetal, 98%