Merck
  • [Synthesis of 11-phenoxyundecyl phosphate and its use as an acceptor substrate in the reaction with UDP-GlcNAc : polyprenyl phosphate GlcNAc-phosphotransferase from Salmonella arizona O:59].
[Synthesis of 11-phenoxyundecyl phosphate and its use as an acceptor substrate in the reaction with UDP-GlcNAc : polyprenyl phosphate GlcNAc-phosphotransferase from Salmonella arizona O:59].
Bioorganicheskaia khimiia (2009-07-22)
L L Danilov, V V Veselovskiĭ, N M Balagurova, T N Druzhinina
ABSTRACT
A new scheme of synthesis of 11-phenoxyundecyl phosphate from 11-bromoundecanoic acid was suggested for its ability to react as an acceptor of 2-acetamido-2-deoxy-alpha-D-glucopyranosyl phosphate in a reaction catalyzed by UDP-N-acetylglucosamine : polyprenyl phosphate N-acetylglucosamine phosphotransferase from Salmonella arizona O:59.
Sigma-Aldrich
11-Bromoundecanoic acid, technical grade, 95%